Learn More
American Research Products Inc Mouse Collagen alpha 1 XVIII
Supplier:  American Research Products Inc CSBE07974M
Encompass
Mouse Collagen alpha-1 (XVIII) chain (COL18A1/ES) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26123