Learn More
American Research Products Inc Mouse Creatine Kinase MM isoen
Supplier:  American Research Products Inc CSBE14407M
Encompass
Mouse Creatine Kinase MM isoenzyme CK-MM ELISA Kit from CUSABIO Detection Range 0 156 mU/mL-10 mU/mL Sensitivity 0 039 mU/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-28858