Learn More
Sino Biological Anti-Cathepsin B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50084RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Cathepsin B / CTSB (Catalog#50084-M08H; P10605; Met1-Phe339). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0200