Learn More
Alpco Ltd Mouse Cystatin C ELISA
SDP
Supplier:  Alpco Ltd 41CYCMSE01
Encompass
The Mouse Cystatin C ELISA is a highly sensitive two-site enzyme linked immunoassay for measuring Cystatin C in mouse biological samples. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2885196