Learn More
Abcam Mouse Cytokeratin 18 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2436781X96TESTS
Encompass_Preferred
Mouse Cytokeratin 18 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Cytokeratin 18 with a sensitivity of 27 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1875220