Learn More
Raybiotech Inc Quantibody Mouse Cytokine Array 7 Kit.
Supplier:  Raybiotech Inc QAMCYT72
Encompass_Preferred
Detects 40 Mouse Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: BAFF R, Betacellulin (BTC), C5a, CCL6, CD48 (SLAMF2), CD6, CD80 (B7-1), Chemerin, Clusterin, Cystatin C, DAN, DLL4, EDAR, Endocan, Fetuin A, H60, IL-33 (IL-1 F11), IL-7 R alpha, Kremen-1, Limitin, Lipocalin-2 (NGAL), LOX-1, Lungkine (CXCL15), Marapsin, MBL-2, Meteorin, Nope, NOV (CCN3), Osteoactivin (GPNMB), OX40 Ligand, P-Cadherin, Periostin, PlGF-2, Progranulin, Prostasin, Renin 1, Testican 3, TIM-1 (KIM-1), TRAIL (TNFSF10), Tryptase epsilon
Catalog No. 50-195-0880