Learn More
American Research Products Inc Mouse D Dimer D2D ELISA Kit
Supplier:  American Research Products Inc CSBE13584M
Encompass
Mouse D-Dimer D2D ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26850