Learn More
American Research Products Inc Mouse Disintegrin and metallop
Supplier:  American Research Products Inc CSBEL001277MO
Encompass
Mouse Disintegrin and metalloproteinase domain-containing protein 17 (ADAM17) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27550