Learn More
Biomatik Corporation Mouse Dopa Decarboxylase (DDC) ELISA Kit
Supplier:  Biomatik Corporation EKU12241
Encompass_Preferred
Mouse Dopa Decarboxylase (DDC) ELISA Kit for detecting DDC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88533. Gene ID: 13195. Target Synonyms: AADC; Aromatic L-Amino Acid Decarboxylase; DOPA decarboxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7997