Learn More
Biomatik Corporation Mouse Doublecortin (DCX) ELISA Kit
Supplier:  Biomatik Corporation EKU03790
Encompass_Preferred
Mouse Doublecortin (DCX) ELISA Kit for detecting DCX in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88809. Gene ID: 13193. Target Synonyms: DBCN; DC; LISX; SCLH; XLIS; Doublin; Doublecortex; Lissencephalin-X; Lissencephaly,X-Linked; Neuronal migration protein doublecortin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6163