Learn More
American Research Products Inc Mouse Ectonucleotide pyrophosp
Supplier:  American Research Products Inc CSBEL007679MO
Mouse Ectonucleotide pyrophosphatase/phosphodiesterase family member 1 (ENPP1) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27919