Learn More
Lonza Walkersville Mouse Embryonic Fibroblasts (MEF)
Supplier:  Lonza Walkersville MFB481
Encompass
Cryopreserved Mouse Primary Embryonic Fibroblasts (MEF) are dissociated from CD-1 mouse embryos, expanded by culture and then cryopreserved.Cells are guaranteed for 5 population doublings, and display morphologic and growth properties equivalent to fresh cells. Clonetics MEF are convenient and easy to use, allowing the researcher to focus on results.Recommended Media: Lonza's DMEM High Glucose w/ L-Gln, 500 mL (Part 12-604F) containing 10% FBS (Sigma-Aldrich Part F6178)Benefits:Ready-To-Use No tissue dissection or animal handling required Increased data reproducibilityCharacteristic morphology and staining tests confirm tissue origin and cell typeNegative for mycoplasma, bacteria, yeast and fungiCertificate of Analysis (CoA) is available for every batch Content:≥ 2 million cryopreserved cells in a 1.0 mL cell suspension
Catalog No. NC0695160