Learn More
Sciencell Research Laboratories Mouse Embryonic Fibroblasts from C57BL/6
Supplier:  Sciencell Research Laboratories M754057
Encompass
MEF from ScienCell Research Laboratories are isolated from embryonic day 14 C57BL/6 mouse embryos. These cells are cryopreserved at P0 and delivered frozen. Each vial contains 5 x 10^6 cells in 1 ml volume.
Catalog No. NC1876935