Learn More
ATCC Mouse ES Cell Basal Medium
Supplier:  ATCC SCRR2011
Encompass
ATCC Mouse ES Cell Basal Medium is a sterile liquid tissue culture medium containing phenol red that has been optimized to support the undifferentiated self-renewal cycle related to mouse embryonic stem cell growth. The medium contains a stable glutamine dipeptide, providing a long shelf-life when handled and stored properly. The medium may be supplemented by the addition of ES-Cell Qualified Fetal Bovine Serum (FBS) or a serum replacement designed for mouse ES cells. This medium can support the growth of mouse ES cells on either a mitotically inactivated layer of mouse embryonic fibroblast (MEF) feeder cells, or in a feeder-free environment using gelatin-coated plates.
Catalog No. 50-238-3794