Learn More
Sino Biological Recombinant Mouse CD23 Protein (His Tag)
Supplier:  Sino Biological 50695M07H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse FCER2A isoform A (P20693-1) (Glu 51-Pro 331) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-3140