Learn More
Quidel Corporation MicroVue Mouse FGF-23 (C-Term) ELISA Kit
SDP
Supplier:  Quidel Corporation 606300
Encompass
The MicroVue Mouse FGF-23 (C-Term) ELISA Kit is a for the measurement of FGF-23 in plasma or cell culture media.
Catalog No. NC9965267