Learn More
Sino Biological Mouse FGFR3 Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEK500715
Encompass_Preferred
The Mouse FGFR3 / CD333 ELISA Pair Set is for the quantitative determination of Mouse FGFR3 / CD333.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse FGFR3 / CD333 coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse FGFR3 / CD333 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse FGFR3 / CD333 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse FGFR3 / CD333 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86543