Learn More
Abcam Mouse Fibronectin ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2109671X96TESTS
Encompass_Preferred
Mouse Fibronectin ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Fibronectin with a sensitivity of 251 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 5 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1928878