Learn More
American Research Products Inc Mouse gonadotropin releasing h
Supplier:  American Research Products Inc CSBE08152M
Encompass
Mouse gonadotropin-releasing hormone GnRH ELISA Kit from CUSABIO Detection Range 7 8 pg/ml - 500 pg/ml Sensitivity 1 95 pg/ml Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28668