Learn More
Biomatik Corporation Mouse Growth Differentiation Factor 5 (GDF5) CLIA Kit
Supplier:  Biomatik Corporation EKU09863
Encompass_Preferred
Mouse Growth Differentiation Factor 5 (GDF5) CLIA Kit for detecting GDF5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 4.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P43027. Gene ID: 14563. Target Synonyms: CDMP1; LAP4; SYNS2; BMP14; Radotermin; Cartilage-Derived Morphogenetic Protein-1; Bone morphogenetic protein 14; Lipopolysaccharide-associated protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8416