Learn More
American Research Products Inc Mouse Heat shock protein HSP 9
Supplier:  American Research Products Inc CSBEL010808MO
Encompass
Mouse Heat shock protein HSP 90-beta (HSP90AB1) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27343