Learn More
STEMCELL Technologies Mouse Hepatic Organoids, Size: 1 Unit
SDP
Supplier:  STEMCELL Technologies 70932
Encompass
Cryopreserved Mouse Hepatic Organoids provide a convenient way to establish or standardize hepatic organoid cultures in the laboratory. Each vial contains mouse hepatic progenitor organoids (derived from the liver of a C57BL/6 mouse) that were cultured in HepatiCult™ Organoid Growth Medium (Mouse; Catalog #06030) and cryopreserved in CryoStor CS10 (Catalog #07930). Using cryopreserved Mouse Hepatic Organoids enables establishment of hepatic organoid cultures without the need to isolate hepatic ducts from primary tissue, eliminating the need for access to fresh mouse tissue. The organoids can be passaged and expanded using HepatiCult™ Organoid Growth Medium (Mouse) and refrozen in CryoStor CS10. Mouse hepatic progenitor organoid cultures can be used for research in a variety of fields, including liver biology and disease modeling, cancer, and toxicity assay development.
Catalog No. NC1471925
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov