Learn More
Diagnostic Biosystems MOUSE HRP KIT UnoVue™ Mouse HRP Kit
Supplier:  Diagnostic Biosystems UM100PD
Encompass_Preferred
The UnoVue Plus Mouse HRP/DAB Detection System is a polymer-based 1-step horseradish peroxidase-labeled detection system for immunohistochemistry. This kit can detect mouse IgG and IgM antibodies, with significantly lower background than biotin/avidin detection systems. The Mouse UnoVue Plus HRP/DAB Detection System can be used for manual staining or with any automated staining instrument. The working solution of Stable DAB/Plus is stable for a week, offering convenience and cost savings due to less hazardous waste generation. This species-specific detection system is particularly useful for multiplex immunohistochemical staining assays.\nKit Contents: Peroxidase Block, Anti-Mouse HRP Polymer, Stable DAB/Plus Buffer, Stable DAB/Plus Chromogen, and an empty mixing bottle for Stable DAB/Plus.
Catalog No. 50-240-7337