Learn More
American Research Products Inc Mouse hypoXia inducible factor
Supplier:  American Research Products Inc CSBE08541M
Encompass
Mouse hypoXia-inducible factor 1a HIF-1a ELISA Kit from CUSABIO Detection Range 0 625 pg/mL-40 pg/mL Sensitivity 0 156 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29681