Learn More
American Research Products Inc Mouse ICOS ligand ICOSLG ELI
Supplier:  American Research Products Inc CSBEL010958MO
Encompass
Mouse ICOS ligand (ICOSLG) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27694