Learn More
Sino Biological Mouse IFN-γ ELISpot KIT (ALP) 10 Kit (96 Tests)
Supplier:  Sino Biological EK50709A10
Encompass_Preferred
The kit applies for the detection of mouse IFN-γ at the cellular level by counting IFN-γ-secreting cells.
Catalog No. 50-006-96268