Learn More
Abcam Mouse IgE ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1577181X96TESTS
Encompass_Preferred
Mouse IgE ELISA Kit ab157718 is used to quantify Mouse IgE with a sensitivity of 1.83 ng/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1588924