Learn More
MOLECULAR DEPOT LLC Mouse IgG - 50 mg
Supplier:  MOLECULAR DEPOT LLC A2010033
Encompass_Preferred
Mouse IgG Purified consists of mouse IgG antibodies affinity-purified on protein A Sepharose The product can be used as non specific blockers in immunoassays ELISA WB and immunohistochemistry Readily available in bulk amounts and at bulk prices Biotinylation and other labeling services are also available For in vitro research use only
Catalog No. 50-000-13382