Learn More
Immunology Consultants Mouse IgG ELISA Kit - (Min. Reactivity With Bv, Ho, Hu)
SDP
Supplier:  Immunology Consultants E90GXSPP
Encompass_Preferred
The Mouse IgG test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgG in mouse biological samples. For the quantitative determination of total mouse IgG in biological samples that may contain bovine,horse or human proteins mmunoglobulins.
Catalog No. 50-240-6616