Learn More
Rockland Immunochemicals Mouse IgG (H&L) Antibody Biotin Conjugated Pre-Adsorbed 1 mg
SDP
Supplier:  Rockland Immunochemicals 610706124
Encompass_Preferred
Mouse IgG antibody generated in donkey detects specifically Mouse IgG whole molecule. This secondary antibody anti-Mouse is ideal for investigators who routinely perform ELISA Sandwich ELISA titration assays western-blot immunoprecipitation and more generally immunoassays.
Catalog No. 50-194-3912