Learn More
Rockland Immunochemicals Mouse IgG IgA IgM (H&L) Antibody Alkaline Phosphatase Conjugated 1 mg
SDP
Supplier:  Rockland Immunochemicals 610105130
Encompass_Preferred
Anti-Mouse IgG IgA IgM Alkaline Phosphatase Conjugated Antibody generated in goat detects reactivity to Mouse IgG Mouse IgA and Mouse IgM. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.
Catalog No. 50-194-3898