Learn More
STEMCELL Technologies Mouse IgG1, kappa Isotype Control Antibody, Clone MOPC-21, Size: 500 µg
SDP
Supplier:  STEMCELL Technologies 60070
Encompass
Clone MOPC-21 is a mouse monoclonal IgG1 antibody suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody. Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used. If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen. This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5376