Learn More
American Research Products Inc Mouse IgG2a Fc Silenced
Supplier:  American Research Products Inc IVMB05531MG
Encompass
Mouse IgG2a Fc Silenced (D265A) Isotype Control Purified No Carrier Protein is a isotype control produced in mouse Available in 1mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-04788