Learn More
Abcam Mouse IgG1 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1330451X96TESTS
Mouse IgG1 ELISA Kit ab133045 is used to quantify Mouse IgG1 with a sensitivity of 3 ng/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1880969