Learn More
Meso Scale Discovery V-PLEX Mouse IL-17A/F Kit, 96-well, 5 Plates, IL-17A/F, Mouse, LLOD 0.231 pg/mL, Dynamic Range 0.231 - 1620 pg/mL, Serum, Plasma, Cell Culture Supernatant, Urine

Supplier: Meso Scale Discovery K152WND2
The MESO SCALE DISCOVERY V-PLEX Mouse IL-17A/F Kit is a validated immunoassay for quantitatively measuring mouse IL-17A/F, featuring a high sensitivity and broad dynamic range. This 96-well kit ensures accurate and reproducible results across various sample types, making it ideal for research in immunology, inflammation, and cytokine studies.
- Platform: V-PLEX
- Analyte: IL-17A/F
- Species reactivity: Mouse
- Kit size: 5 plates
- Plate type: 96-well
- Lower limit of detection (LLOD): 0.231 pg/mL
- Dynamic range: 0.231 - 1,620 pg/mL
- Sample types: Serum, Plasma, Cell Culture Supernatant, Urine
- Capture antibody type: Rat Monoclonal
- Detection antibody type: Rat Monoclonal
- Recombinant standards expressed in: E. coli
- Recommended instruments: MESO SECTOR S 600MM, MESO QuickPlex SQ 120MM
- Usage statement: For Research Use Only
- Storage condition: Multi-component
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.