Learn More
Abcam Mouse IL-18 Matched Antibody Pair Kit, 5 x 96 Tests
Supplier:  Abcam AB2188085X96TESTS
Encompass_Preferred
Mouse IL-18 Matched Antibody Pair Kit is a kit for ELISA development for the measurement of Mouse IL-18 by sandwich ELISA method.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1882107