Learn More
Sino Biological Mouse IL-2R gamma/CD132 Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEK500875
Encompass_Preferred
The Mouse IL2RG / CD132 ELISA Pair Set is for the quantitative determination of Mouse IL2RG / CD132.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for IL2RG / CD132 coated on a 96-well plate. Standards and samples are added to the wells, and any IL2RG / CD132 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-IL2RG / CD132 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of IL2RG / CD132 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86624