Learn More
BIOHIPPO Mouse IL11Ra Microsample
SDP
Supplier:  BIOHIPPO BHE15210945
Encompass
Mouse IL11Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL11Ra in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 062 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse IL11Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse IL11Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse IL11Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15166