Learn More
Sino Biological Mouse IL6ST ELISA Pair Set 5PP (Sino Biological)
Supplier:  Sino Biological SEK501355
Encompass_Preferred
Each vial contains 60 ng of recombinant mouse IL6ST. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 750 pg/mL is recommended.
Catalog No. 50-237-2834