Learn More
American Research Products Inc Mouse Immunoglobulin D IgD E
Supplier:  American Research Products Inc CSBE15761M
Encompass
Mouse Immunoglobulin D (IgD) ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-30258