Learn More
Raybiotech Inc C-Series Mouse Inflammation Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAMINF12
Encompass_Preferred
Detects 40 Mouse Inflammatory Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: BLC (CXCL13), CD30 Ligand (TNFSF8), Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), Fas Ligand (TNFSF6), Fractalkine (CX3CL1), GCSF, GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-3, IL-4, IL-6, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17A, I-TAC (CXCL11), KC (CXCL1), Leptin, LIX, Lymphotactin (XCL1), MCP-1 (CCL2), M-CSF, MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 gamma, RANTES (CCL5), SDF-1 alpha (CXCL12 alpha), I-309 (TCA-3/CCL1), TECK (CCL25), TIMP-1, TIMP-2, TNF alpha, TNF RI (TNFRSF1A), TNF RII (TNFRSF1B)
Catalog No. 50-194-8740