Learn More
Mercodia Inc MOUSE INSULIN ELISA KIT (10X 9
SDP
Supplier:  Mercodia Inc 10124710
Encompass_Preferred
Mercodia Mouse Insulin ELISA provides a method for the quantitative determination of insulin in mouse serum, plasma, or cell culture media. The Mercodia Mouse Insulin ELISA is an easy-to-use assay based on highly specific monoclonal antibodies with insignificant or no cross-reactivity to C-peptide or proinsulin.
Catalog No. NC9856617