Learn More
Mercodia Inc MOUSE INSULIN ELISA KIT
Supplier: Mercodia Inc 10124701
Mercodia Mouse Insulin ELISA provides a method for the quantitative determination of insulin in mouse serum, plasma, or cell culture media. The Mercodia Mouse Insulin ELISA is an easy-to-use assay based on highly specific monoclonal antibodies with insignificant or no cross-reactivity to C-peptide or proinsulin.