Learn More
American Research Products Inc Mouse Insulin like growth fact
Supplier:  American Research Products Inc CSBE04581M
Encompass
Mouse Insulin-like growth factor 1 IGF-1 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 151 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26622