Learn More
American Research Products Inc Mouse intercellular adhesion m
Supplier:  American Research Products Inc CSBE04575MIS
Encompass
Mouse intercellular adhesion molecule 1 ICAM-1 ELISA KIT from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 15 6 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26423