Learn More
PBL Assay Science Mouse Interferon Alpha neutralizing viral protein
Supplier:  PBL Assay Science 121852
Encompass
Viral C12R IFN Antagonist selectively neutralizes all mouse IFN alpha subtypes
Catalog No. NC2047772