Learn More
Abcam Mouse Interferon beta protein, 100UG
Supplier:  Abcam AB282390100UG
Encompass_Preferred
Mouse Interferon beta protein is a Mouse Full Length protein, in the 30 to 379 aa range, expressed in HEK 293, <= 0.005 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC, MS.
Protein/ Peptide/ Enzyme Synonyms: Ifb, Ifnb, Ifnb1, Interferon beta, IFN-beta
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4006