Learn More
Raybiotech Inc G-Series Mouse Interleukin Array 2 Kit.
Supplier:  Raybiotech Inc GSMINT21
Encompass_Preferred
Semi-quantitative measurement of 10 mouse interleukins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IL-1 ra (IL-1 F3), IL-2 R alpha, IL-6 R, IL-11, IL-12 p40, IL-17B, IL-17E (IL-25), IL-17F, IL-20, IL-28A (IFN-lambda 2)
Catalog No. 50-195-0470