Learn More
STEMCELL Technologies Mouse Intestinal Organoids, Size: 1 Unit
SDP
Supplier:  STEMCELL Technologies 70931
Encompass
Cryopreserved Mouse Intestinal Organoids provide a convenient way to establish or standardize intestinal organoid cultures in your laboratory. Each vial contains 200 mouse intestinal organoids derived from the small intestine of C57BL/6 mice that were cultured in IntestiCult™ Organoid Growth Medium (Mouse) and cryopreserved in CryoStor CS10. Using cryopreserved Mouse Intestinal Organoids enables establishment of intestinal organoid cultures without the need to isolate intestinal crypts from primary tissue, eliminating the need for access to fresh mouse tissue and making it easy to standardize experimental starting materials. The organoids can be passaged and expanded using IntestiCult™ Organoid Growth Medium (Mouse) and refrozen in CryoStor CS10. Mouse intestinal organoid cultures can be used for research in a variety of fields, including epithelial cell biology, cancer, cystic fibrosis, microbiomics, intestinal immunology, and bacterial or viral pathogenesis.
Catalog No. 50-197-5868
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov