Learn More
XenoTech,LLC M1000.IS9(NP) Pooled CD-1 Mouse Intestinal S9 Fraction, PMSF-Free, 1.0 mL
SDP
Supplier:  XenoTech,LLC M1000.IS9(NP)
Encompass_Preferred
The SEKISUI XENOTECH LLC M1000.IS9(NP) is a pooled CD-1 mouse intestinal S9 fraction, prepared without PMSF to maintain high esterase activity. This product is ideal for in vitro metabolism studies, particularly for evaluating esterase enzymes' roles in drug metabolism. It offers a comprehensive mix of enzymes, including cytochromes P450 and UGTs, assuring superior quality and consistency, differentiating it from other offerings in terms of enzyme content and preparation methods.
Catalog No. NC3312008